Order Entry
Germany
ContactUsLinkComponent
Sodium hydroxide 10.0 M NaOH (10.0N) volumetric solution, Fluka™
Corrosive
Sodium hydroxide 10.0 M NaOH (10.0N) volumetric solution, Fluka™
Catalog # 656054-1L
CAS Number:  
Corrosive
Sodium hydroxide 10.0 M NaOH (10.0N) volumetric solution, Fluka™
Catalog # 656054-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    9.80 - 10.20 N
  • Carbonate (as Na₂CO₃):
    Max. 0.12 N
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:120 °C
Density:1.350 g/cm³
Storage Temperature:Ambient
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548