

HMDS (1,1,1,3,3,3-hexamethyldisilazane), Semiconductor grade (Honeywell 17713)
Catalog # HONC40215H-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay (GC):Min. 98 %
- Density (D 20/4):0.770 - 0.775
- Refractive index (n 20/D):1.4070 - 1.4090
- Non-volatile matter:Max. 5 ppm
- Aluminium (Al):Max. 0.1 ppm
- Arsenic, antimony (as As):Max. 0.01 ppm
- Calcium (Ca):Max. 0.1 ppm
- Copper (Cu):Max. 0.1 ppm
- Iron (Fe):Max. 0.1 ppm
- Sodium (Na):Max. 0.5 ppm
- Lead (Pb):Max. 0.05 ppm
- Zinc (Zn):Max. 0.1 ppm
- Chloride (Cl):Max. 5 ppm
Specification Test Results
About this item
:
:161.39 g/mol
Boiling Pt:126 °C (1.013 hPa)
Melting Pt:-80 °C
Melting Pt:–80 °C
Density:0.770…0.775 g/cm³ (20 °C)
Flash Pt:25.9 %(V)
Storage Temperature:Ambient
EINECS:213-668-5
UN:3286
ADR:3,II
:GHS02
:GHS06
CAS Number:999-97-3
Formula:(CH₃)₃SiNHSi(CH₃)₃
MDL Number:MFCD00008259