Zinc acetate dihydrate ≥98% ACS, Fluka™
Catalog # HONC383058-25KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Drum with inliner
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Identity (IR):complying
- Assay:98.0 - 101.0 %
- pH (5 %, 25 °C):6.0 - 7.0
- in CH₃COOH insoluble matters:Max. 0.005 %
- Calcium (Ca):Max. 0.005 %
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 0.01 %
- Magnesium (Mg):Max. 0.005 %
- Sodium (Na):Max. 0.05 %
- Lead (Pb):Max. 0.002 %
- Chloride (Cl):Max. 5 ppm
- Sulfate (SO₄):Max. 0.005 %
- Appearance (color):complying
- Appearance (form):complying
Specification Test Results
About this item
:
:219.51 g/mol
Merck Index:12,10256
Melting Pt:100 °C
Density:1.750 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-170-2
UN:3077
ADR:9,III
:GHS07
:GHS05
CAS Number:5970-45-6
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961