FAM benzine, 65-95 °C DIN 51635, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64742-49-0
:
MDL Number:MFCD00081849
Boiling Pt:65…95 °C
Melting Pt:–30 °C
Density:0.76 g/cm³ (15 °C)
Flash Pt:–26 °C
UN:3295
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Boiling range:60 - 95 °C
- Density (15 °C):0.695 - 0.710 g/cm³
- Amount of distillate 90 °C:85 - 95 Vol.%
- Residue from evaporation (DIN EN ISO 6246):Max. 0.03 g/l
- n-Hexane (GC):Max. 5 %
- Benzene (GC):Max. 0.1 %
- APHA:Max. 10