Order Entry
Germany
ContactUsLinkComponent
ESD cleanroom shoe covers, anti-slip
ESD cleanroom shoe covers, anti-slip
Catalog # HOIT110228
undefined
ESD cleanroom shoe covers, anti-slip
Catalog # HOIT110228

Specifications

  • Garment application:
    Cleanroom
  • Colour:
    Blue
  • Length:
    457 mm
  • Pk:
    300

Specifications

About this item

The anti-static ESD shoe covers are manufactured with fabric strips.

  • Skid-free soles for extra traction and improved safety
  • Made of PP
  • Non sterile

Weight: 37 g/m²

Packaging: 100 pieces/bag, 3 bags/case.