Order Entry
Germany
ContactUsLinkComponent
Acetone ≥99.5%, AR®, Macron Fine Chemicals™
Acetone ≥99.5%, AR®, Macron Fine Chemicals™
   Sustainable Options Available
Certificates
undefined
Acetone ≥99.5%, AR®, Macron Fine Chemicals™
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay:
    ≥99.5%
  • Water (KF):
    ≤0.3%
  • Titrable Acid (meq/g):
    ≤0.0005
  • Residue after Evaporation:
    ≤0.001%
  • Alcohols:
    ≤0.06%
  • Substances Insoluble in Water:
    Passes Test
  • Substances Reducing KMnO₄ (as O):
    ≤0.0002%

Specification Test Results

Related Information