SYPRO® Ruby protein blot stain
Catalog # LONZ50565
Supplier: LONZA
CAS Number:
Specifications
- Pk:200 mL
- Description:SYPRO® Ruby Protein Blot Stain
- Seq:006
Specifications
About this item
SYPRO™ Ruby is 60-times more sensitive than reversible protein blot stains like Ponceau S, and 30-times more sensitive than Amido Black or Coomassie® brilliant blue stains.
- Visualize proteins prior to transfer - does not interfere with protein activity
- Versatile - Excellent for staining native proteins
- Safe - No acids or organic solvents necessary
- Sensitive - Detects as little as 4-8 ng protein per band