Overalls, PP
About this item
Ideal in decorating, general dust and dirt protection, factory visits, farming, food industry, general industrial handling, laboratory, painting.
- High-tech fabric that offers cloth-like comfort that is easy to wear
- Offers excellent protection against a wide range of hazards and nuisances
Weight: 22 to 55 g/m²
Weight: 22 to 55 g/m²
Certifications: Certified ISO 9001, ISO 13485, EN 340, EN 149.
2 Options Available Below
Product Details & Documents
Ideal in decorating, general dust and dirt protection, factory visits, farming, food industry, general industrial handling, laboratory, painting.
- High-tech fabric that offers cloth-like comfort that is easy to wear
- Offers excellent protection against a wide range of hazards and nuisances
Weight: 22 to 55 g/m²
Weight: 22 to 55 g/m²
Certifications: Certified ISO 9001, ISO 13485, EN 340, EN 149.
Specifications for Product Family
Specifications
- Catalog No:
- HOIT120919
- HOIT129491
- Garment application:
- Chemical protection|Food
- Chemical protection|Food
Specifications
Product Family Options
With hoods
Product Information
- ColourClothing SizeAvailabilityPrice
- HOIT120919Overalls with hood zipper, 58 g/m²Specifications:
More
Specifications
Cat. No.HOIT120919Garment applicationChemical protection|Food
With collars
Product Information
- ColourClothing SizeAvailabilityPrice
- HOIT129491Overalls with collar, non-woven, 30 g/m²Specifications:
More
Specifications
Cat. No.HOIT129491Garment applicationChemical protection|Food
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



