Order Entry
Germany
ContactUsLinkComponent
VWR®, Cork Plate, Square
VWR®, Cork Plate, Square
Catalog # 217-0895
Supplier:  VWR Collection
VWR®, Cork Plate, Square
Catalog # 217-0895
Supplier:  VWR Collection

Specifications

  • Pk:
    20

Specifications

About this item

  • Thickness: 8 mm
  • Length: 300 mm