Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
NucSpot®
NucSpot® Live 650 1000X in DMSO live cell nuclear stain
NucSpot® Live 650 1000X in DMSO live cell nuclear stain
NucSpot® Live 650 1000X in DMSO live cell nuclear stain
Supplier:
Biotium
Certificates
Expand image
NucSpot® Live 650 1000X in DMSO live cell nuclear stain
Supplier:
Biotium
About this item
:
Storage Temperature:
Refrigerator