Nucleosil 120 C18, HPLC Columns
: Avantor
Packed with classical spherical porous silica available in a wide range of pore sizes. The 100 Å and 120 Å have similar relative retention characteristics, however the 100 Å has the greater capacity whilst the 120 Å exhibits a lower backpressure.
- Good mechanical stability
- Good batch to batch reproducibility
Frequently Bought Together