Order Entry
Germany
ContactUsLinkComponent
Health
3,3'-Diaminobenzidine (DAB), tablets buffer substance, for microscopical staining, Sigma-Aldrich®
Catalog # 1.02924.0001
Supplier:  Merck
CAS Number:  
Health
3,3'-Diaminobenzidine (DAB), tablets buffer substance, for microscopical staining, Sigma-Aldrich®
Catalog # 1.02924.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 Tab.
  • Seq:
    0935

Specifications

Specification Test Results

  • Suitability for microscopy:
    passes test

Specification Test Results

About this item

 :
 :214.27 g/mol
 :GHS08
Storage Temperature:Refrigerator
 :1
CAS Number:91-95-2
Formula:C₁₂H₁₄N₄
MDL Number:MFCD00007725