Order Entry
Germany
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, 8-Methoxypsoralen
 :  Merck
 
undefined
Supelco® Organic Reference Standard, 8-Methoxypsoralen
 :  Merck

 

  • CAS No.:
    298-81-7
  • Storage conditions:
    RT
  • Hazard class description:
    H302 - H317 - H341 - H351
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00005009
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    8-MOP; 9-Methoxyfuro[3,2-g][1]benzopyran-7-one; Ammoidin; Methoxsalen; Xanthotoxin
  • Melting point:
    148 - 150 °C (lit.)
  • Molecular weight:
    216,19