Supelco® Organic Reference Standard, 20α-Hydroxycholesterol
H6378-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:516-72-3
- Description:20α-Hydroxycholesterol
- Storage conditions:RT
- Shipping conditions:STD
- MDL No.:MFCD16661190
- M-Clarity quality level:MQ200
- Synonym(s):5-Cholestene-3β,20α-diol
- Molecular weight:402,65