Potassium fluoride solution, NMR reference standard, Supelco®
Catalog # 612871-10X8IN
Supplier: Merck
CAS Number:
Specifications
- Concentration:0.2 M in water, D2O 10 % (99.9 atom % D)
- Pk:10
- Standards:NMR reference standard
- Description:Potassium fluoride solution
- Description:Potassium fluoride solution
- Pk Info:NMR tube size 8 mm x 8 in.