Order Entry
Germany
ContactUsLinkComponent
1,2-Dichlorobenzene solution, NMR reference standard, Supelco®
1,2-Dichlorobenzene solution, NMR reference standard, Supelco®
Catalog # 612502-10X8IN
Supplier:  Merck
CAS Number:  
undefined
1,2-Dichlorobenzene solution, NMR reference standard, Supelco®
Catalog # 612502-10X8IN
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1% in acetone-d6 (99.9 atom % D)
  • Pk:
    10
  • Standards:
    NMR reference standard
  • Description:
    1,2-Dichlorobenzene solution
  • Description:
    1,2-Dichlorobenzene solution
  • Pk Info:
    NMR tube size 3 mm x 8 in.

Specifications

About this item