Order Entry
Germany
ContactUsLinkComponent
Tetramethylsilane solution, NMR reference standard, Supelco®
Tetramethylsilane solution, NMR reference standard, Supelco®
Catalog # 551333-1EA
Supplier:  Merck
CAS Number:  
undefined
Tetramethylsilane solution, NMR reference standard, Supelco®
Catalog # 551333-1EA
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    12% in chloroform
  • Pk:
    1
  • Standards:
    NMR reference standard
  • Description:
    Tetramethylsilane solution
  • Description:
    Tetramethylsilane solution
  • Pk Info:
    NMR tube size 5 mm x 8 in.

Specifications

About this item