Order Entry
Germany
ContactUsLinkComponent
 
Acetic acid glacial ≥99.5%, AR®, Macron Fine Chemicals™
 
undefined
Acetic acid glacial ≥99.5%, AR®, Macron Fine Chemicals™
 

 

  • Assay:
    99.5%
  • Acetaldehyde (CH₃CHO):
    ≤0.05%
  • Residue after Evaporation:
    ≤0.002%
  • Substances Reducing KMnO₄ (as HCOOH):
    ≤0.02%
  • Chloride (Cl):
    ≤0.001%
  • Sulfate (SO₄):
    ≤0.001%
  • Heavy Metals (as Pb):
    ≤0.0002%