Order Entry
Germany
ContactUsLinkComponent
Flammable
Health
Irritant
Congo red staining kit
Catalog # 1.01641.0001
Supplier:  Merck
CAS Number:  
undefined
Flammable
Health
Irritant
Congo red staining kit
Catalog # 1.01641.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre case
  • Pk:
    0,196 L
  • Seq:
    5022
  • Pk Info:
    Preparation capacity 50 slides

Specifications

Specification Test Results

  • Suitability for microscopy:
    passes test
  • Amyloid:
    pink to red; in polarized ligt green metachromasis
  • Nuclei:
    dark blue
  • Connective tissue:
    light red

Specification Test Results

About this item

 :
 :696.67 g/mol
Storage Temperature:Ambient
UN:3316
ADR:9,II
 :GHS02
 :GHS08
 :GHS07
 :1
CAS Number:573-58-0
Formula:C₃₂H₂₂N₆Na₂O₆S₂
MDL Number:MFCD00004028