Order Entry
Germany
ContactUsLinkComponent
 
Corrosive
Irritant
Masson-Goldner staining kit for histology, Sigma-Aldrich®
Catalog # 1.00485.0001
Supplier:  Merck
CAS Number:  
undefined
Corrosive
Irritant
Masson-Goldner staining kit for histology, Sigma-Aldrich®
Catalog # 1.00485.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    4 x 500 ml (Preparation Capacity 400 – 500 slides)

Specifications

Specification Test Results

  • Suitability for microscopy (tissue section):
    Passes test
  • Nuclei:
    Dark brown to black
  • Cytoplasm:
    Brick red
  • Musculature (muscles):
    Bright red
  • Connective tissue:
    Green
  • Acid mucosubstances:
    Green
  • Erythrocytes:
    Bright orange

Specification Test Results

About this item

 :
UN:2790
ADR:8,III
 :GHS05
 :GHS07
 :1