

Haematoxylin modified solution Gill II used in nuclear staining, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:517-28-2
:208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111
:302.28 g/mol
Density:1.04…1.06 g/cm3 (20 °C)
:GHS08
:GHS07
Storage Temperature:Ambient
:1
Specification Test Results
- Suitability for microscopy:passes test
- Nuclei (Vaginal smear):blue to dark violet
- Cyanophilic cytoplasm:bluish green to green
- Eosinophilic cytoplasm:pink