Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, Withanolide A
Supelco® Organic Reference Standard, Withanolide A
Catalog # 74776-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Withanolide A
Catalog # 74776-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    32911-62-9
  • Description:
    Withanolide A
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD07779143
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (5α,6α,7α,22R)-6,7-Epoxy-5,20,22-trihydroxy-1-oxo-ergosta-2,24-dien-26-oic acid δ-lactone; Withaniol
  • Molecular weight:
    470,60

Specifications

About this item