Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, Violaxanthin
Supelco® Organic Reference Standard, Violaxanthin
Catalog # 91904-1MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Violaxanthin
Catalog # 91904-1MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Standards:
    Analytical standard
  • CAS No.:
    126-29-4
  • Description:
    Violaxanthin
  • Storage conditions:
    –70 °C
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3S,3′S,5R,5′R,6S,6′S)-5,6:5′,6′-Diepoxy-5,5′,6,6′-tetrahydro-β,β-carotene-3,3′-diol
  • Molecular weight:
    600,87

Specifications

About this item