Order Entry
Germany
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Verbascose
Catalog # 56217-5MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Verbascose
Catalog # 56217-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    546-62-3
  • Description:
    Verbascose
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD03095487
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    α-ᴅ-Galp-(1→6)-α-ᴅ-Galp-(1→6)-α-ᴅ-Galp-(1→6)-α-ᴅ-Glup(1↔2)-β-ᴅ-Fruf
  • Molecular weight:
    828,72

Specifications

About this item