Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, Sesamin
Catalog # 59867-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Sesamin
Catalog # 59867-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    607-80-7
  • Description:
    Sesamin
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H317
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00216134
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Fagarol
  • Molecular weight:
    354,35

Specifications

About this item

Caution: P261 - P272 - P280 - P302 + P352 - P333 + P313 - P362 + P364