Order Entry
Germany
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, Sennoside A

Supelco® Organic Reference Standard, Sennoside A

Supplier:   Merck
Supelco® Organic Reference Standard, Sennoside A
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, Sennoside A
Supelco® Organic Reference Standard, Sennoside A

2 Options Available Below

Product Details & Documents

Caution: P264 - P270 - P301 + P312 - P501

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 68909-5MG-F
  • 68909-25MG-F
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • CAS No.:
  • 81-27-6
  • 81-27-6
  • MDL No.:
  • MFCD00151527
  • MFCD00151527
  • Formula:
  • C₄₂H₃₈O₂₀
  • C₄₂H₃₈O₂₀
  • Molecular weight:
  • 862,74
  • 862,74
  • Hazard class description:
  • H302
  • H302
  • Flash point:
  • Not applicable
  • Not applicable
  • Shipping conditions:
  • STD
  • STD
  • Storage conditions:
  • 2...30 °C
  • 2...30 °C

Specifications

Product Family Options

Product Information

  • Standards
    Form
    Pk
    Availability
    Price
  • 68909-5MG-F
    Sennoside A
    Sennoside A
    Analytical standard
    Neat
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 129,00 €
    129,00 €
    1 * 5 mg
       129,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    68909-5MG-F
    M-Clarity quality level
    MQ100
    CAS No.
    81-27-6
    MDL No.
    MFCD00151527
    Formula
    C₄₂H₃₈O₂₀
    Molecular weight
    862,74
    Hazard class description
    H302
    Flash point
    Not applicable
    Shipping conditions
    STD
    Storage conditions
    2...30 °C
  • 68909-25MG-F
    Sennoside A
    Sennoside A
    Analytical standard
    Neat
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 509,00 €
    509,00 €
    1 * 25 mg
       509,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    68909-25MG-F
    M-Clarity quality level
    MQ100
    CAS No.
    81-27-6
    MDL No.
    MFCD00151527
    Formula
    C₄₂H₃₈O₂₀
    Molecular weight
    862,74
    Hazard class description
    H302
    Flash point
    Not applicable
    Shipping conditions
    STD
    Storage conditions
    2...30 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.