Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, Secologanin
Supelco® Organic Reference Standard, Secologanin
Catalog # 50741-5MG-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Secologanin
Catalog # 50741-5MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    19351-63-4
  • Description:
    Secologanin
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD16661163
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    388,37

Specifications

About this item