Supelco® Organic Reference Standard, Piperitone
Artikelnr. 79899-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:89-81-6
- Description:Piperitone
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319
- Shipping conditions:STD
- MDL No.:MFCD00045532
- M-Clarity quality level:MQ100
- Synonym(s):3-Carvomenthenone; 6-Isopropyl-3-methyl-2-cyclohexen-1-one; p-Menth-1-en-3-one
- Molecular weight:152,23
- Flash point:83,5 °C - Closed cup
Specifications
About this item
- GHS signal: GHS07
Caution: P302 + P352 - P305 + P351 + P338