Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, Hederacoside C
Supelco® Organic Reference Standard, Hederacoside C
Catalog # 97151-50MG-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Hederacoside C
Catalog # 97151-50MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    14216-03-6
  • Description:
    Hederacoside C
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00198062
  • M-Clarity quality level:
    MQ100
  • Melting point:
    222 °C (dec.) (lit.)
  • Molecular weight:
    1221,38

Specifications

About this item

  • GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501