Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, Erythrodiol
Supelco® Organic Reference Standard, Erythrodiol
Catalog # 09258-10MG-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Erythrodiol
Catalog # 09258-10MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    545-48-2
  • Description:
    Erythrodiol
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00017294
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Olean-12-ene-3β,28-diol
  • Melting point:
    230...231 °C (lit.)
  • Formula:
    C₃₀H₅₀O₂
  • Molecular weight:
    442,72
  • Flash point:
    Not applicable

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.