Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, Bisphenol S, analytical standard, Supelco®
Organic reference standard, Bisphenol S, analytical standard, Supelco®
Catalog # 43034-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Bisphenol S, analytical standard, Supelco®
Catalog # 43034-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    80-09-1
  • Description:
    Bisphenol S, ≥98,0% (HPLC)
  • Melting point:
    245...250 °C (lit.)
  • Formula:
    C₁₂H₁₀O₄S 
  • Description:
    Bisphenol S
  • Form:
    Neat

Specifications

About this item