Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, Bisphenol F bis(3-chloro-2-hydroxypropyl) ether, mixture of 3 isomers ortho-ortho, ortho-para, para-para, Supelco®
Organic reference standard, Bisphenol F bis(3-chloro-2-hydroxypropyl) ether, mixture of 3 isomers ortho-ortho, ortho-para, para-para, Supelco®
Catalog # 15139-250MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Bisphenol F bis(3-chloro-2-hydroxypropyl) ether, mixture of 3 isomers ortho-ortho, ortho-para, para-para, Supelco®
Catalog # 15139-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Description:
    Bisphenol F bis(3-chloro-2-hydroxypropyl) ether, mixture of 3 isomers ortho-ortho, ortho-para, para-para, ≥90% (mixture of isomers, HPLC)
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₉H₂₂Cl₂O₄
  • Description:
    Bisphenol F bis(3-chloro-2-hydroxypropyl) ether
  • Form:
    Neat

Specifications

About this item

Bisphenol F bis(3-chloro-2-hydroxypropyl) ether a additive used during synthesis of organosol resin preventing theremal degradation of polymer. During the removal of hydrochloric acid in organosol resin results in this chlorinated compound.

  • Used as reference standard in RP-HPLC-APCI-MS-MS analysis for optimising and quantifying pressurised liquid extraction for BFDGE and its derivatives from canned food

Standard for determining toxic monomers released from polymers of the inner coating of cans.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.