Organic reference standard, Benzylidene camphor sulphonic acid, analytical standard, Supelco®
Catalog # 97085-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:56039-58-8
- Description:Benzylidene camphor sulfonic acid, ≥97,0% (HPLC)
- Formula:C₁₇H₂₀O₄S
- Description:Benzylidene camphor sulfonic acid
Specifications
About this item
Benzylidene camphor sulphonic acid (BCSA) is an organic ultraviolet absorbing compound.
- Used primarily in cosmetics (sunscreens, lipsticks, shampoos and hair sprays)