Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1-Kestose
Supelco® Organic Reference Standard, 1-Kestose
Catalog # 72555-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 1-Kestose
Catalog # 72555-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    470-69-9
  • Description:
    1-Kestose
  • Seq:
    02
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00142647
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    β-ᴅ-Fruf-(2→1)-β-ᴅ-Fruf-(2→1)-α-ᴅ-Glup
  • Molecular weight:
    504,44

Specifications

About this item