Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-Isopulegol
Supelco® Organic Reference Standard, (−)-Isopulegol
Catalog # 59770-5ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (−)-Isopulegol
Catalog # 59770-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    89-79-2
  • Description:
    (−)-Isopulegol
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00134655
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (1R,2S,5R)-2-Isopropenyl-5-methylcyclohexanol; (1R,3R,4S)-p-Menth-8-en-3-ol
  • Molecular weight:
    154,25
  • Density:
    0,912 g/ml at 25 °C (lit.)
  • Flash point:
    78 °C - Closed cup
  • Boiling point:
    212 °C (lit.)

Specifications

About this item

  • GHS signal: GHS07
Caution: P261 - P264 - P270 - P301 + P312 - P302 + P352 - P305 + P351 + P338