Order Entry
Germany
ContactUsLinkComponent
Zearalenone solution, certified reference material, Supelco®
Zearalenone solution, certified reference material, Supelco®
Catalog # CRM46916
Supplier:  Merck
CAS Number:  
undefined
Zearalenone solution, certified reference material, Supelco®
Catalog # CRM46916
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    50 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    Zearalenone solution
  • Description:
    Zearalenone solution
  • Pk Info:
    Ampoule of 1 ml

Specifications

About this item