ContactUsLinkComponent
VWR®, Cleanroom Gloves, Nitrile, Sterile, ISO 5
VWR®, Cleanroom Gloves, Nitrile, Sterile, ISO 5
Supplier:  VWR Collection
Certificates
undefined
VWR®, Cleanroom Gloves, Nitrile, Sterile, ISO 5
Supplier:  VWR Collection
Certificates
Specifications
  • Catalog No:
  • 112-5429
  • 112-5428
  • 112-5427
  • 112-5431
  • 112-5426
  • 112-5430
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Cleanroom class:
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • Finish:
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,08 - 0,12
  • 0,08 - 0,12
  • 0,08 - 0,12
  • 0,08 - 0,12
  • 0,08 - 0,12
  • 0,08 - 0,12
  • Finger thickness:
  • 0,12 - 0,18
  • 0,12 - 0,18
  • 0,12 - 0,18
  • 0,12 - 0,18
  • 0,12 - 0,18
  • 0,12 - 0,18
  • Palm thickness:
  • 0,10 - 0,15
  • 0,10 - 0,15
  • 0,10 - 0,15
  • 0,10 - 0,15
  • 0,10 - 0,15
  • 0,10 - 0,15
Specifications