Order Entry
Germany
ContactUsLinkComponent
ASTM® D5307 crude oil internal standard, certified reference material, Supelco®
ASTM® D5307 crude oil internal standard, certified reference material, Supelco®
Catalog # 48479.
Supplier:  Merck
CAS Number:  
ASTM® D5307 crude oil internal standard, certified reference material, Supelco®
Catalog # 48479.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    (equal weights of the hydrocarbons listed)
  • Pk:
    1
  • Description:
    ASTM(R) D5307 Crude oil internal standard

Specifications

About this item

ASTM® D5307 crude oil internal standard has been used in the quantitative determination of diamondoid compounds like adamantane in specific organic solvents like dichloromethane, hexane, carbon-tetrachloride, hexane , heptadecane, hexadecane, pentadecane and tetradecane, using infrared attenuated total reflection spectroscopy (IR-ATR).

Analytes:

Heptadecane 25% (w/w); hexadecane 25% (w/w); pentadecane 25% (w/w); tetradecane 25% (w/w)