Trifluoroacetic anhydride acylation reagent, for GC derivatisation
Supplier: MACHEREY-NAGEL
Certificates
About this item
CAS Number:407-25-0
:
Formula:C₄F₆O₃
MDL Number:MFCD00000416
:210.03 g/mol
Boiling Pt:39.5–40.5 °C (760 mm Hg)
Density:1.490 g/cm³ (d20°/4°)
UN:3316
ADR:9,II
:GHS05
:GHS07