Order Entry
Germany
ContactUsLinkComponent
 
 
 
 
Chlorotrimethylsilane ≥99.0% (by GC), liquid, LiChropur™ for GC derivatisation, Supelco®
 :  Merck
 
undefined
 
 
 
Chlorotrimethylsilane ≥99.0% (by GC), liquid, LiChropur™ for GC derivatisation, Supelco®
 :  Merck
 

 

  • Appearance (Color):
    Colorless
  • Purity (GC Area %):
    ≥ 99.0 %
  • Proton NMR Spectrum:
    Conforms to Structure