Pharmaceutical analytical standard, Marbofloxacin, reference material, Honeywell Fluka™
HONCR1645-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:115550-35-1
- Description:Marbofloxacin
- Formula:C₁₇H₁₉FN₄O₄
- Storage temperature:Ambient
- Description:Marbofloxacin
- Form:Powder
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.