ContactUsLinkComponent
Overalls, PP/PE with hood
Overalls, PP/PE with hood
Catalog # 113-9303
Supplier:  VWR Collection
undefined
Overalls, PP/PE with hood
Catalog # 113-9303
Supplier:  VWR Collection
Specifications
  • Clothing Size:
    M
  • Garment application:
    General Purpose,
    Industrial
  • Colour:
    White
  • Seq:
    02
  • Pk:
    50
  • Weight:
    40 g/m²
Specifications
About this item
Overalls made of PE with laminated PP.
  • With hood
  • Elasticated waist, wrist and ankles
  • Zip fastening in front
  • Latex-free
Weight: 40 g/m²
Colour: White/blue.
Packaging: One piece per bag and 50 pieces per carton.