Supelco® Organic Reference Standard, EPA 552.2 Haloacetic Acids Mix
49107-U
: Merck
:
- Concentration:2000 μg/ml each component in methyl tert-butyl ether
- Pk:1
- Standards:Certified reference material
- Description:EPA 552.2 Haloacetic acids mix
- Storage conditions:2…8 °C
- Hazard class description:H225 - H315
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:–33 °C
- GHS signal: GHS02, GHS07
: P210 - P233 - P240 - P241 - P242 - P303 + P361 + P353