Organic reference standard, Methyl hexacosanoate, analytical standard, Supelco®
Catalog # 52203-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:5802-82-4
- Description:Methyl hexacosanoate
- M-Clarity quality level:MQ100
- Melting point:63...66 °C
- Formula:C₂₇H₅₄O₂
- Storage temperature:2…8 °C
- Description:Methyl hexacosanoate
Specifications
About this item
Methyl hexacosanoate is a compound, which can be isolated from lingwu bituminous coal.
- Bottomless glass bottle
- Contents are inside inserted fused cone