Organic reference standard, Methyl heneicosanoate, analytical standard, Supelco®
51535-1G
: Merck
:
- Pk:1 g
- Standards:Analytical standard
- CAS No.:6064-90-0
- Description:Methyl heneicosanoate
- M-Clarity quality level:MQ100
- Melting point:48...50 °C (lit.)
- Formula:C₂₂H₄₄O₂
- Storage temperature:2…8 °C
- Description:Methyl heneicosanoate
- Boiling point:207 °C/4 mmHg (lit.)
Methyl heneicosanoate can find applications as an internal standard in the quantitative determination of fatty acids and their isomers composition using gas chromatography method of analysis (GC).