Supelco® PESTANAL® Organic Reference Standard, Guazatine Acetate Salt
Catalog # 37915-100MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:115044-19-4
- Description:Guazatine acetate salt
- Storage conditions:2...30 °C
- Hazard class description:H302 + H312 - H330 - H400
- Shipping conditions:STD
- MDL No.:MFCD01724079
- M-Clarity quality level:MQ100
- Formula:C₂₀H₄₅N₇O₂
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P260 - P273 - P280 - P301 + P312 - P302 + P352 + P312 - P304 + P340 + P310