Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, Glycidamide
Catalog # 04704-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Glycidamide
Catalog # 04704-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    5694-00-8
  • Description:
    Glycidamide
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302 - H315 - H317 - H319 - H335 - H350
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00468053
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2,3-Epoxypropanamide; Oxiranecarboxamide
  • Formula:
    C₃H₅NO₂
  • Molecular weight:
    87,08
  • Storage temperature:
    2...8°C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS08, GHS07
Caution: P201 - P280 - P301 + P312 + P330 - P302 + P352 - P305 + P351 + P338 - P308 + P313