Supelco® PESTANAL® Organic Reference Standard, Furilazole
Catalog # 32431-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:121776-33-8
- Description:Furilazole
- Storage conditions:2...8 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD03792863
- M-Clarity quality level:MQ100
- Synonym(s):(RS)-3-Dichloroacetyl-5-(2-furanyl)-2,2-dimethyl-1,3-oxazolidine
- Formula:C₁₁H₁₃Cl₂NO₃
- Molecular weight:278,13
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07