Supelco® PESTANAL® Organic Reference Standard, Diquat Dibromide Monohydrate
Catalog # 45422-250MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:6385-62-2
- Description:Diquat dibromide monohydrate
- Storage conditions:2...8 °C
- Hazard class description:H301 + H311 - H315 - H317 - H319 - H330 - H335 - H372 - H410
- Shipping conditions:STD
- MDL No.:MFCD00211319
- M-Clarity quality level:MQ100
- Synonym(s):Diquat
- Formula:C₁₂H₁₂Br₂N₂ ·H₂O
- Molecular weight:362,06
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
Caution: P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310 - P305 + P351 + P338 - P314