Tape dispenser, metal, Stericlin®
Catalog # VEPA3FSZB410302
Supplier: VP Medical Packaging
Specifications
- For Use With:Tapes with 19 - 25 mm width
- Colour:Blue
- Description:Metal tape dispenser, desk type
- Pk:1
Specifications
About this item
Blue metal dispenser for use with Stericlin® adhesive tapes.